missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylene cyanol FF, ∽75%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 440700100

 View more versions of this product

Catalog No. AC440700100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylene cyanol FF
∽75%
MFCD00019481
VVLFAAMTGMGYBS-KRQUPCAFSA-M
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
538.61
Blue/Gray to Green
Powder
2650-17-1
C25H27N2NaO6S2
Acid blue 147 monosidium salt, C.I. 42135
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
538.61
Glass bottle
10 g

Chemical Identifiers

2650-17-1
538.61
VVLFAAMTGMGYBS-KRQUPCAFSA-M
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
C25H27N2NaO6S2
MFCD00019481
Acid blue 147 monosidium salt, C.I. 42135
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only