missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylene Cyanol FF 85%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 422690250

 View more versions of this product

Catalog No. AC422690250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylene cyanol
4463-44-9
C25H28N2NaO7S2
C.I. 43535, Cyanol FF
Authentic
4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium
131854036
Extra Pure
Blue to Red
Crystalline Powder
85%
80% min.
MFCD00019481
JKPDSWLBCLGXEQ-UHFFFAOYSA-N
CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]
555.616
554.61
Glass bottle
25 g

Chemical Identifiers

4463-44-9
555.616
JKPDSWLBCLGXEQ-UHFFFAOYSA-N
131854036
CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na]
C25H28N2NaO7S2
MFCD00019481
C.I. 43535, Cyanol FF
4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 224-728-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only