Learn More
Xylene Cyanol, pure
CAS: 4463-44-9 | C25H28N2NaO7S2 | 555.616 g/mol
$88.82 - $3767.38
Chemical Identifiers
| CAS | 4463-44-9 |
|---|---|
| Molecular Formula | C25H28N2NaO7S2 |
| Molecular Weight (g/mol) | 555.616 |
| MDL Number | MFCD00019481 |
| InChI Key | JKPDSWLBCLGXEQ-UHFFFAOYSA-N |
| Synonym | C.I. 43535, Cyanol FF |
| PubChem CID | 131854036 |
| IUPAC Name | 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium |
| SMILES | CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC422690050
|
Thermo Scientific Chemicals
422690050 |
5 g | Glass bottle |
Each for $88.82
|
|
||||
|
AC422690250
|
Thermo Scientific Chemicals
422690250 |
25 g | Glass bottle |
Each for $323.15
|
|
||||
|
AC422691000
|
Thermo Scientific Chemicals
422691000 |
100 g | Glass bottle |
Each for $905.83
|
|
||||
|
AC422695000
|
Thermo Scientific Chemicals
422695000 |
500 g | Glass bottle |
Each for $3,767.38
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4463-44-9 | |
| 555.616 | |
| JKPDSWLBCLGXEQ-UHFFFAOYSA-N | |
| 131854036 | |
| CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na] |
| C25H28N2NaO7S2 | |
| MFCD00019481 | |
| C.I. 43535, Cyanol FF | |
| 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium |
Specifications
| 4463-44-9 | |
| C25H28N2NaO7S2 | |
| C.I. 43535, Cyanol FF | |
| Authentic | |
| 4-[[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonic acid;sodium | |
| 131854036 | |
| Extra Pure | |
| Blue to Red | |
| Crystalline Powder |
| 80% min. | |
| MFCD00019481 | |
| JKPDSWLBCLGXEQ-UHFFFAOYSA-N | |
| CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)O)O)C.[Na] | |
| 555.616 | |
| 554.61 | |
| Glass bottle | |
| 5 g | |
| Xylene cyanol, 85% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 224-728-5
RUO – Research Use Only