missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Plastic Open-Head Drums
High-density polyethylene containers for transport of hazardous solids or liquids
Supplier: Youngstown Barrel & Drum 2346
Description
- Made of chemical-resistant high-density polyethylene
- Open-head drums for transport of hazardous solids
- 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
Specifications
Pail | |
Screw Lid | |
29.14 L | |
For Packaging Small Amounts of Liquid Products | |
50 | |
Screw Lid | |
14 in. | |
11.6 lb. |
UN 1H2/Y41/S | |
7.7 gal. | |
White | |
Plastic | |
Open | |
16.14 in. | |
5.51 cm | |
5.3 Kg |