Youngstown Barrel & Drum Plastic Open-Head Drums
High-density polyethylene containers for transport of hazardous solids or liquids
$49.08 - $234.05
Specifications
Material Plastic
Freight Class 50
Head Style Open
View More Specs
Products 5
Catalog Number Mfr. No. Certifications/Compliance Color Capacity (English) For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Color Capacity (English) For Use With (Application) Price Quantity  
18-940
View Documents
Youngstown Barrel & Drum
1720
UN 1H2/Y100/S and X65/S Blue 14 gal. For Transport of Hazardous Solids or Liquids
Each for $176.43
19-130-4695
View Documents
Youngstown Barrel & Drum
2378
UN 1H2/Y1.5/30 & UN 1H2/Y30/S White 5 gal. For Packaging Small Amounts of Liquid Products
Each for $49.08
19-130-4850
View Documents
Youngstown Barrel & Drum
1556
UN 1H2/X120/S and UN 1H2/Y180/S Blue 30 gal. For Transport of Hazardous Solids or Liquids
Each for $234.05
18-120-570
View Documents
Youngstown Barrel & Drum
1748
UN 1H2/Y54/S White 10.7 gal. For Transport of Hazardous Solids or Liquids
Each for $86.35
18-120-571
View Documents
Youngstown Barrel & Drum
2346
UN 1H2/Y41/S White 7.7 gal. For Packaging Small Amounts of Liquid Products
Each for $113.07
Description
Description
  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
Specifications
Specifications
Plastic
50
Open
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'