Youngstown Barrel & Drum Plastic Open-Head Drums
High-density polyethylene containers for transport of hazardous solids or liquids
Supplier:  Youngstown Barrel & Drum 2378
 View more versions of this product
Catalog No. 19-130-4695
Explore available promotions
Description
Description
  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
Specifications
Specifications
Pail
Screw Lid with Neoprene Gasket
18.92 L
For Packaging Small Amounts of Liquid Products
50
Screw Lid with Neoprene Gasket and Push/Pull Spout
42.54 cm
12.47 in.
31.67 mm
1.98 Kg
UN 1H2/Y1.5/30 & UN 1H2/Y30/S
5 gal.
White
Plastic
Open
16.75 in.
12.03 in.
30.55 cm
4.37 lb.