missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Plastic Open-Head Drums
High-density polyethylene containers for transport of hazardous solids or liquids
Supplier: Youngstown Barrel & Drum 1556
Description
- Made of chemical-resistant high-density polyethylene
- Open-head drums for transport of hazardous solids
- 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
Specifications
Drum | |
Lever Lock Ring | |
114 L | |
For Transport of Hazardous Solids or Liquids | |
50 | |
26.37 in. | |
19 in. | |
15 lb. |
UN 1H2/X120/S and UN 1H2/Y180/S | |
30 gal. | |
Blue | |
Plastic | |
Open | |
66.99 cm | |
48.26 cm | |
6.8 Kg |