missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Plastic Open Head Pails with Screw Lids
Seamless construction prevents leaks
$16.56 - $66.29
Specifications
Closure Type | Screw Lid with Neoprene Gasket |
---|---|
Color | White |
For Use With (Application) | For Packaging Small Amounts of Liquid Products |
Material | Plastic |
Freight Class | 50 |
Catalog Number | Mfr. No. | Capacity (English) | Certifications/Compliance | Includes | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Certifications/Compliance | Includes | Price | Quantity | |||
18-120-240
|
Youngstown Barrel & Drum
2379BH |
3.5 gal. | UN 1H2/Y19/S | Screw Lid with Neoprene Gasket and BIOHAZARD Label |
Each for $41.16
|
|
|||
18-120-241
|
Youngstown Barrel & Drum
2369BH |
6.5 gal. | UN 1H2/Y30/S | Screw Lid with Neoprene Gasket and BIOHAZARD Label |
Each for $56.75
|
|
|||
18-120-242
|
Youngstown Barrel & Drum
2375BH |
5 gal. | UN 1H2/X11.5/S and UN 1H2/Y30/S | Screw Lid with Neoprene Gasket and BIOHAZARD Label |
Each for $56.64
|
|
|||
19-171-855
|
Youngstown Barrel & Drum
2369NG |
6.5 gal. | UN 1H2/Y30/S | Screw Lid with Neoprene Gasket |
Each for $66.29
|
|
|||
19-171-856
|
Youngstown Barrel & Drum
2375NG |
5 gal. | UN 1H2/X11.5/S and UN 1H2/Y30/S | Screw Lid with Neoprene Gasket |
Each for $52.91
|
|
|||
19-171-857
|
Youngstown Barrel & Drum
2423NG |
1.25 gal. | UN 1H2/Y6/S | Screw Lid with Neoprene Gasket |
Each for $16.56
|
|
|||
Description
- Blow-molded, high-density plastic
- Drop handle is comfortable for carrying loaded pail
- Handle folds down for storage or stacking
- White
Specifications
Screw Lid with Neoprene Gasket | |
For Packaging Small Amounts of Liquid Products | |
50 |
White | |
Plastic | |
Open |