missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate, 99%, pure

CAS: 557-34-6 | C4H6O4Zn | 183.48 g/mol

Supplier:  Thermo Scientific Chemicals 370081000

 View more versions of this product

Catalog No. AC370081000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc acetate
557-34-6
Powder
98.5% min. (Complexometry)
(CH3CO2)2Zn
15, 10327
DJWUNCQRNNEAKC-UHFFFAOYSA-L
zinc;diacetate
11192
183.48
Pure
1% max.
83°C to 86°C
Gray-Green to Yellow
100 g
C4H6O4Zn
MFCD00012454
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
CC(=O)[O-].CC(=O)[O-].[Zn+2]
183.48
CHEBI:62984
99%
Authentic
Glass bottle

Chemical Identifiers

557-34-6
183.48
DJWUNCQRNNEAKC-UHFFFAOYSA-L
11192
zinc;diacetate
C4H6O4Zn
MFCD00012454
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
CHEBI:62984
CC(=O)[O-].CC(=O)[O-].[Zn+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiti

GHS Signal Word: Danger

EINECSNumber : 209-170-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only