missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc acetate, 99%, pure, Thermo Scientific Chemicals

$187.67 - $503.18

Chemical Identifiers

CAS 557-34-6
Molecular Formula C4H6O4Zn
Molecular Weight (g/mol) 183.48
MDL Number MFCD00012454
InChI Key DJWUNCQRNNEAKC-UHFFFAOYSA-L
Synonym zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
PubChem CID 11192
ChEBI CHEBI:62984
IUPAC Name zinc;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370080250
SDS View Documents
Thermo Scientific Chemicals
370080250
25 g Glass bottle
Each for $187.67
 
AC370081000
SDS View Documents
Thermo Scientific Chemicals
370081000
100 g Glass bottle
Each for $503.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

557-34-6
183.48
DJWUNCQRNNEAKC-UHFFFAOYSA-L
11192
zinc;diacetate
C4H6O4Zn
MFCD00012454
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
CHEBI:62984
CC(=O)[O-].CC(=O)[O-].[Zn+2]

Specifications

83°C to 86°C
Gray-Green to Yellow
25 g
C4H6O4Zn
MFCD00012454
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
CC(=O)[O-].CC(=O)[O-].[Zn+2]
183.48
CHEBI:62984
99%
Authentic
Glass bottle
557-34-6
Powder
98.5% min. (Complexometry)
(CH3CO2)2Zn
15, 10327
DJWUNCQRNNEAKC-UHFFFAOYSA-L
zinc;diacetate
11192
183.48
Pure
1% max.
Zinc acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiti

GHS Signal Word: Danger

EINECSNumber : 209-170-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only