missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc fluoride, 98%, Thermo Scientific Chemicals

$495.14 - $495.14

Chemical Identifiers

CAS 7783-49-5
Molecular Formula F2Zn
Molecular Weight (g/mol) 103.38
MDL Number MFCD00011298
InChI Key BHHYHSUAOQUXJK-UHFFFAOYSA-L
Synonym zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
PubChem CID 10129912
IUPAC Name zinc;difluoride
SMILES [F-].[F-].[Zn+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC363912500
SDS View Documents
Thermo Scientific Chemicals
363912500
250 g Plastic bottle
Each for $495.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-49-5
103.38
BHHYHSUAOQUXJK-UHFFFAOYSA-L
10129912
[F-].[F-].[Zn+2]
F2Zn
MFCD00011298
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
zinc;difluoride

Specifications

1500.0°C
7783-49-5
Crystals or Powder
98%
ZnF2
06,514
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride
BHHYHSUAOQUXJK-UHFFFAOYSA-L
zinc;difluoride
10129912
98%
Zinc fluoride, 98%
872.0°C to 910.0°C
Brown to White
250 g
F2Zn
MFCD00011298
15, 10335
Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl, hno3 and nh4oh, insoluble in alcohol
[F-].[F-].[Zn+2]
103.38
103.38
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

EINECSNumber : 232-001-9

RTECSNumber : ZH3500000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only