Learn More
Zinc iodide, 98+%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 208062500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc iodide | |
98+% | |
Beige-Yellow to White | |
250 g | |
I2Zn | |
MFCD00011299 | |
15, 10339 | |
319.19 | |
≥98% | |
Glass bottle | |
4.7400g/mL |
446°C | |
10139-47-6 | |
Hygroscopic Crystalline Powder or Crystals | |
98% min. (Argentometry) | |
ZnI2 | |
01,1294; 05,765; 10,462; 11,604; 12,576; 13,88 | |
Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether | |
319.19 | |
Pure | |
4.74 |
Chemical Identifiers
10139-47-6 | |
I2Zn | |
319.19 | |
MFCD00011299 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 233-396-0
TSCA : TSCA
RUO – Research Use Only