Learn More
Zinc iodide, 98+%, pure, Thermo Scientific Chemicals
$67.31 - $879.69
Chemical Identifiers
CAS | 10139-47-6 |
---|---|
Molecular Formula | I2Zn |
Molecular Weight (g/mol) | 319.19 |
MDL Number | MFCD00011299 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC208060500
|
Thermo Scientific Chemicals
208060500 |
50 g | Glass bottle |
Each for $67.31
|
|
||||
AC208062500
|
Thermo Scientific Chemicals
208062500 |
250 g | Glass bottle |
Each for $261.81
|
|
||||
AC208060010
|
Thermo Scientific Chemicals
208060010 |
1 kg | Glass bottle |
Each for $879.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10139-47-6 | |
I2Zn | |
319.19 | |
MFCD00011299 |
Specifications
446°C | |
Beige-Yellow to White | |
50 g | |
I2Zn | |
MFCD00011299 | |
15, 10339 | |
319.19 | |
≥98% | |
Glass bottle | |
4.7400g/mL |
10139-47-6 | |
Hygroscopic Crystalline Powder or Crystals | |
98% min. (Argentometry) | |
ZnI2 | |
01,1294; 05,765; 10,462; 11,604; 12,576; 13,88 | |
Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether | |
319.19 | |
Pure | |
4.74 | |
Zinc iodide, 98+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 233-396-0
TSCA : TSCA
RUO – Research Use Only