missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211660050

 View more versions of this product

Catalog No. AC211660050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc nitrate hexahydrate
10196-18-6
Adhering Crystals
98%
Zn(NO3)2·6H2O
14,10144
Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
297.48
297.46
Extra Pure
36.0°C
White
5 g
H12N2O12Zn
MFCD00149889
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc;dinitrate;hexahydrate
15865313
98%
Glass Bottle

Chemical Identifiers

10196-18-6
297.48
JGPSMWXKRPZZRG-UHFFFAOYSA-N
15865313
H12N2O12Zn
MFCD00149889
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye pr

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only