Learn More
Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals
$35.18 - $233.96
Chemical Identifiers
CAS | 10196-18-6 |
---|---|
Molecular Formula | H12N2O12Zn |
Molecular Weight (g/mol) | 297.48 |
MDL Number | MFCD00149889 |
InChI Key | JGPSMWXKRPZZRG-UHFFFAOYSA-N |
Synonym | zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a |
PubChem CID | 15865313 |
SMILES | O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC211660050
|
Thermo Scientific Chemicals
211660050 |
5 g | Glass Bottle |
Each for $35.18
|
|
||||
AC211662500
|
Thermo Scientific Chemicals
211662500 |
250 g | Plastic bottle |
Each for $88.24
|
|
||||
AC211660010
|
Thermo Scientific Chemicals
211660010 |
1 kg | Plastic bottle |
Each for $233.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10196-18-6 | |
297.48 | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
15865313 |
H12N2O12Zn | |
MFCD00149889 | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
36.0°C | |
White | |
5 g | |
H12N2O12Zn | |
MFCD00149889 | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
zinc;dinitrate;hexahydrate | |
15865313 | |
98% | |
Glass Bottle |
10196-18-6 | |
Adhering Crystals | |
98% | |
Zn(NO3)2·6H2O | |
14,10144 | |
Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
297.48 | |
297.46 | |
Extra Pure | |
Zinc nitrate hexahydrate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye pr
GHS Signal Word: Danger
RUO – Research Use Only