missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc nitrate hexahydrate, 99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 012313A3

 View more versions of this product

Catalog No. AA12313A3


Explore available promotions

Description

Description

Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc nitrate hexahydrate
10196-18-6
2 kg
H12N2O12Zn
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
JGPSMWXKRPZZRG-UHFFFAOYSA-N
zinc(2+) hexahydrate dinitrate
15865313
Hygroscopic
∽36°C
Crystalline
99% (metals basis)
MFCD00149889
Soluble in water,alcoholSoluble in water and ethanol.
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
297.48
297.47 (189.38 Anhydrous)
2.07 g/mL

Chemical Identifiers

10196-18-6
297.48
JGPSMWXKRPZZRG-UHFFFAOYSA-N
15865313
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H12N2O12Zn
MFCD00149889
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
zinc(2+) hexahydrate dinitrate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P370+P378q-P501c

H272-H302-H315-H319-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 231-943-8

RTECSNumber : ZH4775000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only