missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc phthalocyanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03955304
Description
Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc phthalocyanine | |
Purple | |
C32H16N8Zn | |
2 g | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] | |
2735172 | |
577.91 | |
Powder |
14320-04-8 | |
Odorless | |
MFCD00041955 | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
577.916 | |
CHEBI:51218 | |
95% |
Chemical Identifiers
14320-04-8 | |
577.916 | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
2735172 | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] |
C32H16N8Zn | |
MFCD00041955 | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
CHEBI:51218 |
Safety and Handling
EINECSNumber : 238-262-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only