missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc phthalocyanine, 95%, Thermo Scientific Chemicals
$61.92 - $712.21
Chemical Identifiers
CAS | 14320-04-8 |
---|---|
Molecular Formula | C32H16N8Zn |
Molecular Weight (g/mol) | 577.916 |
MDL Number | MFCD00041955 |
InChI Key | PODBBOVVOGJETB-UHFFFAOYSA-N |
Synonym | zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % |
PubChem CID | 2735172 |
ChEBI | CHEBI:51218 |
SMILES | C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3955304
|
Thermo Scientific Chemicals
03955304 |
2 g |
Each for $61.92
|
|
|||||
AA3955309
|
Thermo Scientific Chemicals
03955309 |
10 g |
Each for $172.58
|
|
|||||
AA3955318
|
Thermo Scientific Chemicals
03955318 |
50 g |
Each for $712.21
|
|
|||||
Description
Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14320-04-8 | |
577.916 | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
2735172 | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] |
C32H16N8Zn | |
MFCD00041955 | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
CHEBI:51218 |
Specifications
14320-04-8 | |
Odorless | |
MFCD00041955 | |
zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 % | |
PODBBOVVOGJETB-UHFFFAOYSA-N | |
577.916 | |
CHEBI:51218 | |
95% | |
Zinc phthalocyanine |
Purple | |
C32H16N8Zn | |
2 g | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2] | |
2735172 | |
577.91 | |
Powder |
Safety and Handling
EINECSNumber : 238-262-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only