missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc Sulfate Heptahydrate, 99.0-102.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals
Supplier: MP Biomedicals Inc 0219145280
Specifications
Zinc Sulfate Heptahydrate | |
Crystalline | |
99.0-102.0% | |
MFCD00149894 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc(2+) heptahydrate sulfate | |
62640 | |
287.54 |
7446-20-0 | |
100 g | |
H14O11SZn | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
ACS Reagent |
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc(2+) heptahydrate sulfate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Safety and Handling
Recommended Storage : Room Temperature