Learn More
Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03339922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Zinc sulfate | |
heptahydrate | |
Lump | |
99.0-103.0% | |
MFCD00149894 | |
Soluble in water,glycerol. | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
ACS | |
1.97 |
100°C | |
7446-20-0 | |
100 g | |
H14O11SZn | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc;sulfate;heptahydrate | |
62640 | |
287.54 (161.43 Anhydrous) | |
Hygroscopic |
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc;sulfate;heptahydrate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Safety and Handling
P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-793-3
RTECSNumber : ZH5300000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only