missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
$66.82 - $443.77
Chemical Identifiers
CAS | 7446-20-0 |
---|---|
Molecular Formula | H14O11SZn |
Molecular Weight (g/mol) | 287.54 |
MDL Number | MFCD00149894 |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
PubChem CID | 62640 |
ChEBI | CHEBI:32312 |
IUPAC Name | zinc;sulfate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA3339922
|
Thermo Scientific Chemicals
03339922 |
100 g |
Each for $66.82
|
|
||||||
AA3339936
|
Thermo Scientific Chemicals
03339936 |
500 g |
Each for $141.16
|
|
||||||
AA33399A4
|
Thermo Scientific Chemicals
33399A4 |
2.5 kg |
Each for $443.77
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
zinc;sulfate;heptahydrate |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Specifications
100°C | |
Lump | |
99.0-103.0% | |
MFCD00149894 | |
Soluble in water,glycerol. | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
ACS | |
1.97 |
7446-20-0 | |
100 g | |
H14O11SZn | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc;sulfate;heptahydrate | |
62640 | |
287.54 (161.43 Anhydrous) | |
Hygroscopic | |
Zinc sulfate, heptahydrate |
Safety and Handling
P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-793-3
RTECSNumber : ZH5300000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only