missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate heptahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 33399A4

 View more versions of this product

Catalog No. AA33399A4


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Zinc sulfate heptahydrate
For analysis ACS
Lump
99.0 to 103.0%
MFCD00149894
Soluble in water,glycerol.
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
287.54
CHEBI:32312
ACS Reagent
1.97 g/mL
100°C
7446-20-0
2.5 kg
H14O11SZn
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
RZLVQBNCHSJZPX-UHFFFAOYSA-L
zinc;sulfate;heptahydrate
62640
287.54 (161.43 Anhydrous)
Hygroscopic

Chemical Identifiers

7446-20-0
287.54
RZLVQBNCHSJZPX-UHFFFAOYSA-L
62640
zinc;sulfate;heptahydrate
H14O11SZn
MFCD00149894
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
CHEBI:32312
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-793-3

RTECSNumber : ZH5300000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only