missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 389800010

 View more versions of this product

Catalog No. AC389800010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc sulfate monohydrate
7446-19-7
1 kg
O4SZn·H2O
MFCD00149893
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
RNZCSKGULNFAMC-UHFFFAOYSA-L
zinc;sulfate;hydrate
62639
99%
Plastic bottle
White
Powder
98.5% min. (Complexometry)
ZnSO4·H2O
15, 10359
Solubility in water: 350g/L (20°C). Other solubilities: practically insoluble in ethanol
O.[O-]S(=O)(=O)[O-].[Zn+2]
179.47
179.47
Confirmed (Zinc), Confirmed (Sulfate)

Chemical Identifiers

7446-19-7
179.47
RNZCSKGULNFAMC-UHFFFAOYSA-L
62639
O.[O-]S(=O)(=O)[O-].[Zn+2]
O4SZn·H2O
MFCD00149893
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
zinc;sulfate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only