Learn More
Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals
$105.97 - $344.53
Chemical Identifiers
CAS | 7446-19-7 |
---|---|
Molecular Formula | O4SZn·H2O |
Molecular Weight (g/mol) | 179.47 |
MDL Number | MFCD00149893 |
InChI Key | RNZCSKGULNFAMC-UHFFFAOYSA-L |
Synonym | zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate |
PubChem CID | 62639 |
IUPAC Name | zinc;sulfate;hydrate |
SMILES | O.[O-]S(=O)(=O)[O-].[Zn+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC389802500
|
Thermo Scientific Chemicals
389802500 |
250 g | Plastic bottle |
N/A
|
|
|||||
AC389800010
|
Thermo Scientific Chemicals
389800010 |
1 kg | Plastic bottle |
N/A
|
|
|||||
AC389800050
|
Thermo Scientific Chemicals
389800050 |
5 kg | Plastic bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-19-7 | |
179.47 | |
RNZCSKGULNFAMC-UHFFFAOYSA-L | |
62639 | |
O.[O-]S(=O)(=O)[O-].[Zn+2] |
O4SZn·H2O | |
MFCD00149893 | |
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate | |
zinc;sulfate;hydrate |
Specifications
White | |
Powder | |
98.5% min. (Complexometry) | |
ZnSO4·H2O | |
15, 10359 | |
Solubility in water: 350g/L (20°C). Other solubilities: practically insoluble in ethanol | |
O.[O-]S(=O)(=O)[O-].[Zn+2] | |
179.47 | |
179.47 | |
Confirmed (Zinc), Confirmed (Sulfate) | |
Zinc sulfate monohydrate |
7446-19-7 | |
250 g | |
O4SZn·H2O | |
MFCD00149893 | |
zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate | |
RNZCSKGULNFAMC-UHFFFAOYSA-L | |
zinc;sulfate;hydrate | |
62639 | |
99% | |
Plastic bottle |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only