Learn More
Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
$110.66 - $1756.78
Chemical Identifiers
CAS | 15461-27-5 |
---|---|
Molecular Formula | Cl2OZr |
Molecular Weight (g/mol) | 178.12 |
MDL Number | MFCD00149898 |
InChI Key | IPCAPQRVQMIMAN-UHFFFAOYSA-L |
Synonym | zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 |
PubChem CID | 159678 |
SMILES | Cl[Zr](Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8610818
|
Thermo Scientific Chemicals
08610818 |
50 g |
Each for $110.66
|
|
|||||
AA8610830
|
Thermo Scientific Chemicals
08610830 |
250 g |
Each for $485.77
|
|
|||||
AA86108A1
|
Thermo Scientific Chemicals
086108A1 |
1 kg |
Each for $1,756.78
|
|
|||||
Description
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15461-27-5 | |
178.12 | |
IPCAPQRVQMIMAN-UHFFFAOYSA-L | |
159678 |
Cl2OZr | |
MFCD00149898 | |
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 | |
Cl[Zr](Cl)=O |
Specifications
400°C (decomposition) | |
Lump | |
99.9%, metal basis | |
MFCD00149898 | |
Soluble in water and alcohol. | |
Cl[Zr](Cl)=O | |
178.12 | |
178.13 (Anhydrous) | |
1.91 |
15461-27-5 | |
50 g | |
Cl2OZr | |
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402 | |
IPCAPQRVQMIMAN-UHFFFAOYSA-L | |
oxozirconium;octahydrate;dihydrochloride | |
159678 | |
Hygroscopic | |
Zirconium Dichloride Oxide |
Safety and Handling
When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 231-717-9
RTECSNumber : ZH7250000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only