Learn More
Zirconium dinitrate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.
$138.60 - $1793.45
Chemical Identifiers
CAS | 14985-18-3 |
---|---|
Molecular Formula | N2O7Zr |
Molecular Weight (g/mol) | 231.23 |
MDL Number | MFCD00149896 |
InChI Key | JWBWFTANBHYHBC-UHFFFAOYSA-N |
Synonym | zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis |
PubChem CID | 16211674 |
SMILES | [O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4322414
|
Thermo Scientific Chemicals
04322414 |
25 g |
Each for $138.60
|
|
|||||
AA4322422
|
Thermo Scientific Chemicals
04322422 |
100 g |
Each for $414.85
|
|
|||||
AA4322436
|
Thermo Scientific Chemicals
04322436 |
500 g |
Each for $1,793.45
|
|
|||||
Description
Zirconium dinitrate oxide hydrate is used as a precursor in the production of Pb(Zr0.50Ti0.50)O3 ferroelectric thin films and silicaand alumina-doped nanocrystalline ceria/zirconia. It is also used to prepare cerium oxide/zirconium oxide mixed oxide catalyst supports.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14985-18-3 | |
231.23 | |
JWBWFTANBHYHBC-UHFFFAOYSA-N | |
16211674 |
N2O7Zr | |
MFCD00149896 | |
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis | |
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O |
Specifications
14985-18-3 | |
25 g | |
N2O7Zr | |
zirconium dinitrate oxide hydrate, zirconyl nitrate hydrate, zirconium iv dinitrate oxide hydrate, oxozirconiumbis ylium hydrate dinitrate, zirconium iv oxynitrate hydrate, zirconium dinitrate oxide hydrate, puratronic, zirconium iv oxynitrate hydrate, technical grade, zirconium iv oxynitrate hydrate trace metals basis | |
JWBWFTANBHYHBC-UHFFFAOYSA-N | |
(nitrooxy)(oxo)zirconio nitrate | |
16211674 | |
Zirconium dinitrate oxide hydrate |
Crystalline | |
99.9% (metals basis) | |
MFCD00149896 | |
Soluble in alcohol. | |
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O | |
231.23 | |
231.23 (Anhydrous) |
Safety and Handling
P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314
DOTInformation : Hazard Class: 5.1; Packaging Group: III
EINECSNumber : 237-529-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only