missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206410050

 View more versions of this product

Catalog No. AC206410050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium(IV) chloride
98%
White
5 g
Cl4Zr
MFCD00011306
13,16
Zirconium tetrachloride
233.03
98%
2.8
437.0°C
10026-11-6
Fine Powder
98%
ZrCl4
01,1295
15,10374
Solubility in water: reacts. Other solubilities: forming hcl and zro2, soluble in alcohol and ether
233.03
Glass bottle

Chemical Identifiers

10026-11-6
233.03
Zirconium tetrachloride
Cl4Zr
MFCD00011306
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Reacts violently with water.
Corrosive to the respiratory tract.

GHS P Statement
Wear respiratory protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
C

GHS Signal Word: Danger

EINECSNumber : 233-058-2

RTECSNumber : ZH7175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only