missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium Oxide, Approx. 99%, Spectrum™ Chemical
$169.26 - $12318.47
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18614828
|
Spectrum Chemical Mfg Cor
Z1102125GM |
125 g | Poly Bottle |
Each for $169.26
|
|
||||
18614831
|
Spectrum Chemical Mfg Cor
Z1102500GM |
500 g | Poly Bottle |
Each for $412.18
|
|
||||
18614829
|
Spectrum Chemical Mfg Cor
Z110225KG |
2.5 kg | Poly Bottle |
Each for $1,313.76
|
|
||||
18614827
|
Spectrum Chemical Mfg Cor
Z110212KG |
12 kg | Poly Pail |
Each for $3,521.26
|
|
||||
18614830
|
Spectrum Chemical Mfg Cor
Z110245KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1314-23-4 |
Specifications
1314-23-4 | |
125 g | |
Reagent |
1 | |
∼99% | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.