missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 208375000

 View more versions of this product

Catalog No. AC208375000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconyl chloride octahydrate
White to Yellow
500 g
Cl2H18O9Zr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
FUBACIUATZGHAC-UHFFFAOYSA-N
oxozirconium;octahydrate;dihydrochloride
159678
98%
1.91
13520-92-8
Crystalline Powder
97.5 to 102.5% (Argentometry)
ZrOCl2·8H2O
15, 10384
Solubility in water: soluble in water. Other solubilities: freely soluble in alcohol,insoluble in other organic solvents
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
324.26
322.25
Glass bottle
1.9100g/mL

Chemical Identifiers

13520-92-8
324.26
FUBACIUATZGHAC-UHFFFAOYSA-N
159678
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Cl2H18O9Zr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
oxozirconium;octahydrate;dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only