missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals

$51.14 - $653.77

Chemical Identifiers

CAS 13520-92-8
Molecular Formula Cl2H18O9Zr
Molecular Weight (g/mol) 324.26
MDL Number MFCD00149898
InChI Key FUBACIUATZGHAC-UHFFFAOYSA-N
Synonym zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
PubChem CID 159678
IUPAC Name oxozirconium;octahydrate;dihydrochloride
SMILES O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC208370050
SDS View Documents
Thermo Scientific Chemicals
208370050
5 g Glass bottle
Each for $51.14
 
AC208371000
SDS View Documents
Thermo Scientific Chemicals
208371000
100 g Glass bottle
Each for $211.75
 
AC208375000
SDS View Documents
Thermo Scientific Chemicals
208375000
500 g Glass bottle
Each for $653.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13520-92-8
324.26
FUBACIUATZGHAC-UHFFFAOYSA-N
159678
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Cl2H18O9Zr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
oxozirconium;octahydrate;dihydrochloride

Specifications

13520-92-8
Crystalline Powder
97.5 to 102.5% (Argentometry)
ZrOCl2·8H2O
15, 10384
(10% in water) Clear to slightly hazy solution
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
324.26
322.25
Glass bottle
1.9100g/mL
White to Yellow
5 g
Cl2H18O9Zr
MFCD00149898
zirconyl chloride octahydrate, zirconium oxychloride octahydrate, zirconium chloride oxide octahydrate, dichlorooxozirconium octahydrate, zirconium, dichlorooxo-, octahydrate, zirconium, dichlorooxo-, octahydrate 8ci,9ci, cl2h16o9zr, dsstox_cid_29361, dsstox_rid_83477, dsstox_gsid_49402
FUBACIUATZGHAC-UHFFFAOYSA-N
oxozirconium;octahydrate;dihydrochloride
159678
98%
1.91
Zirconyl chloride octahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only