missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Glass Burette with PTFE Key Stopcock

Glass Burettes with PTFE Key Stopcock are designed with a single straight bore and features easy to read, printed graduations. Class-A and Class-B versions available.

$62.44 - $95.79

Specifications

Autoclavable Autoclavable
Color Clear
For Use With (Equipment) Clamps, lab stand
Material Borosilicate Glass (3.3)
For Use With (Application) Titration
View More Specs

Products 6
Catalog Number Mfr. No. Capacity (Metric) Class Scale Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Class Scale Price Quantity  
S27992
View Documents
Eisco™
CH0240C
50 mL Class A Blue
Each for $63.21
 
S15484
View Documents
Eisco™
CH0240G
50 mL Class B Blue
Each for $85.76
 
S29407
View Documents
Eisco™
CH0240BWT
25 mL Class A White
Each for $62.44
 
S29408
View Documents
Eisco™
CH0240F
25 mL Class B Blue
Each for $65.53
 
S15482
View Documents
Eisco™
CH0240B
25 mL Class A Blue
Each for $74.21
 
S13964
View Documents
Eisco™
CH0240D
100 mL Class A Blue
Each for $95.79
 
Description

Description

  • Made from high quality 3.3 borosilicate
  • Easy to read, chemical resistant ceramic graduations
  • Accurate to DIN ISO 385 standards
  • Straight, single bore
  • Includes interchangeable, self-lubricating PTFE key stopcock
Specifications

Specifications

Autoclavable
Clamps, lab stand
Titration
PTFE
Straight Bore Buret
Clear
Borosilicate Glass (3.3)
PTFE Stopcock
1 year limited manufacturer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'