Learn More
1-Bromobutane, 99%
CAS: 109-65-9 | C4H9Br | 137.02 g/mol
$50.49 - $383.32
Chemical Identifiers
| CAS | 109-65-9 |
|---|---|
| Molecular Formula | C4H9Br |
| Molecular Weight (g/mol) | 137.02 |
| MDL Number | MFCD00000260 |
| InChI Key | MPPPKRYCTPRNTB-UHFFFAOYSA-N |
| Synonym | butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 |
| PubChem CID | 8002 |
| IUPAC Name | 1-bromobutane |
| SMILES | CCCCBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC106770500
|
Thermo Scientific Chemicals
106770500 |
50 mL | Glass bottle |
Each for $50.49
|
|
||||
|
AC106772500
|
Thermo Scientific Chemicals
106772500 |
250 mL | Glass bottle |
Each for $117.99
|
|
||||
|
AC106770010
|
Thermo Scientific Chemicals
106770010 |
1 L | Glass bottle |
Each for $250.79
|
|
||||
|
AC106770025
|
Thermo Scientific Chemicals
106770025 |
2.5 L | Glass bottle |
Each for $383.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109-65-9 | |
| 137.02 | |
| MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
| 8002 | |
| CCCCBr |
| C4H9Br | |
| MFCD00000260 | |
| butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
| 1-bromobutane |
Specifications
| 109-65-9 | |
| 100.0 | |
| Colorless to Yellow | |
| 100.0°C to 104.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4380 to 1.4404 | |
| MFCD00000260 | |
| 01,119 | |
| 15,1555 | |
| Solubility in water: 0.608g/L (30°C). Other solubilities: soluble in alcohol,ether,acetone,chloroform,,benzene and ccl4 | |
| CCCCBr | |
| 137.02 | |
| 137.02 | |
| Liquid |
| 98.5 | |
| -112.0°C | |
| 1.2700g/mL | |
| 10°C | |
| 98.5% min. (GC) | |
| C4H9Br | |
| CH3(CH2)3Br | |
| 50 mL | |
| 1.27 | |
| butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831 | |
| MPPPKRYCTPRNTB-UHFFFAOYSA-N | |
| 1-bromobutane | |
| 8002 | |
| 99% | |
| 1-Bromobutane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 203-691-9
RUO – Research Use Only