missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium dihydrogen phosphate, 98+%, for biochemistry, Thermo Scientific™
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
Synonym | ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
IUPAC Name | azanium;dihydrogen phosphate |
SMILES | [NH4+].OP(O)([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC447842500
|
Thermo Scientific Chemicals
447842500 |
250g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC447840010
|
Thermo Scientific Chemicals
447840010 |
1kg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
azanium;dihydrogen phosphate |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
[NH4+].OP(O)([O-])=O |
Specifications
7722-76-1 | |
98+% | |
NH4H2PO4 | |
15,541 | |
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone | |
[NH4+].OP(O)([O-])=O | |
115.03 | |
CHEBI:62982 | |
98+% | |
Plastic bottle |
250g | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03 | |
Biochemistry | |
Ammonium dihydrogen phosphate, for biochemistry |
Safety and Handling
EINECSNumber : 231-764-5
RUO â Research Use Only