missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) 2,4-pentanedionate, 98%, Thermo Scientific Chemicals

$53.08 - $785.70

Chemical Identifiers

CAS 13395-16-9
Molecular Formula C10H14CuO4
Molecular Weight (g/mol) 261.76
MDL Number MFCD00000016
InChI Key QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Synonym Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
IUPAC Name copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1492014
View Documents
Thermo Scientific Chemicals
A1492014
25 g
Each for $53.08
 
AAA1492018
View Documents
Thermo Scientific Chemicals
A1492018
50 g
Each for $88.40
 
AAA1492022
View Documents
Thermo Scientific Chemicals
A1492022
100 g
Each for $132.36
 
AAA1492036
View Documents
Thermo Scientific Chemicals
A1492036
500 g
Each for $447.97
 
AAA149200B
View Documents
Thermo Scientific Chemicals
A149200B
1000 g
Each for $785.70
 
Description

Description

Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) 2,4-pentanedionate is used as a catalyst for carbene transfer reactions, coupling reactions and Michael addition reactions. It is also used as a catalyst for the polymerization of olefins and transesterification reactions as well as a PVC stabilizer. Further, it is used as a curing agent for epoxy resins, acrylic adhesives, silicone rubbers. In addition to this, it is used as a lubricant additive and paint drier.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

13395-16-9
261.76
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
C10H14CuO4
MFCD00000016
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

13395-16-9
160°C (10 mmHg)
C10H14CuO4
25 g
Bis(acetylacetonato)copper(II); Bis(2,4-pentanedionato)copper(II)
QYJPSWYYEKYVEJ-FDGPNNRMSA-L
copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
261.76
Copper(II) 2,4-pentanedionate
∼286°C (decomposition)
Weak
MFCD00000016
4157957
Insoluble in water.
[Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
261.76
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H360-H500

EINECSNumber : 236-477-9

RTECSNumber : GL6520000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only