missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Reusable PFA Low-Form Beakers

Translucent PFE with raised graduation scale

$957.02 - $2708.83

Specifications

Color Translucent
Material PFA
For Use With (Application) For High Temperature and Chemical Resistance
Temperature (English) Max. Operating 68°F
Temperature (Metric) Max. Operating 20°C
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
10-210-670
View Documents Promotion Details
Corning™
1003P50
50 mL
Case of 12 Each for $957.02
 
10-210-671
View Documents Promotion Details
Corning™
1003P100
100 mL
Case of 12 Each for $1,220.56
 
10-210-672
View Documents Promotion Details
Corning™
1003P250
250 mL
Case of 6 Each for $1,126.61
 
10-210-673
View Documents Promotion Details
Corning™
1003P500
500 mL
Case of 6 Each for $1,759.00
 
10-210-674
View Documents Promotion Details
Corning™
1003P1L
1 L
Case of 6 Each for $2,708.83
 
Description

Description

These beakers are translucent with a raised graduation scale. They feature high temperature and chemical resistance.

Specifications

Specifications

Translucent
For High Temperature and Chemical Resistance
20°C
Autoclavable
Yes
Raised Scale
PFA
68°F
Beaker
Reusable
DIN, ASTM Class A and B
Griffin
SDS
missing translation for 'documents'

missing translation for 'documents'