missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanone cyanohydrin, 98%, Thermo Scientific Chemicals

$173.88 - $580.78

Chemical Identifiers

CAS 931-97-5
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00003818
InChI Key ZDBRPNZOTCHLSP-UHFFFAOYSA-N
Synonym cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
PubChem CID 13610
IUPAC Name 1-hydroxycyclohexane-1-carbonitrile
SMILES C1CCC(CC1)(C#N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2669909
View Documents Promotion Details
Thermo Scientific Chemicals
H2669909
10 g
Each for $173.88
 
AAH2669918
View Documents Promotion Details
Thermo Scientific Chemicals
H2669918
50 g
Each for $580.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

931-97-5
125.171
ZDBRPNZOTCHLSP-UHFFFAOYSA-N
13610
C1CCC(CC1)(C#N)O
C7H11NO
MFCD00003818
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
1-hydroxycyclohexane-1-carbonitrile

Specifications

931-97-5
1.031
118°C (244°F)
1.469
10 g
cyclohexanone cyanohydrin, 1-hydroxycyclohexanecarbonitrile, 1-hydroxy-1-cyclohexanecarbonitrile, cyclohexanone, cyanohydrin, 1-cyano-1-hydroxycyclohexane, cyclohexanecarbonitrile, 1-hydroxy, unii-on43942n5p, ccris 4608, 1-hydroxy-cyclohexanecarbonitrile, cyclohexanoncyanhydrin
C1CCC(CC1)(C#N)O
125.171
125.17
Cyclohexanone cyanohydrin
34°C to 36°C
132°C (19 mmHg)
C7H11NO
MFCD00003818
UN3439
ZDBRPNZOTCHLSP-UHFFFAOYSA-N
1-hydroxycyclohexane-1-carbonitrile
13610
98%
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P501c

H300+H310+H330-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 213-246-0

RTECSNumber : GU7710000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only