missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
CAS: 50-99-7 | C6H12O6 | 180.16 g/mol
$103.25 - $344.89
Chemical Identifiers
| CAS | 50-99-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
| Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
| PubChem CID | 107526 |
| ChEBI | CHEBI:42758 |
| IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC388190010
|
Thermo Scientific Chemicals
388190010 |
1 kg |
Each for $103.25
|
|
|||||
|
AC388190050
|
Thermo Scientific Chemicals
388190050 |
5 kg |
Each for $344.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 50-99-7 | |
| 180.16 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| 107526 | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
| 50-99-7 | |
| 0.5% max. (1 g, 105°C, 16 h) | |
| 1 kg | |
| + 52.75 (25.00°C c=10, H2O, NH4OH) | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 107526 | |
| 180.16 | |
| 0.5% max. (total impurities) |
| 146.0°C | |
| C6H12O6 | |
| 15,4495 | |
| + 52.75 | |
| Solubility in water: soluble. | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.16 | |
| CHEBI:42758 | |
| 97.5 to 102.0% (Anhydrous substance) | |
| D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP |
Safety and Handling
EINECSNumber : 200-075-1
RUO – Research Use Only