Learn More
Dimethoxymethane, 99.5+%
CAS: 109-87-5 | C3H8O2 | 76.09 g/mol
$130.60 - $261.65
Identifiants chimiques
| CAS | 109-87-5 |
|---|---|
| Molecular Formula | C3H8O2 |
| Molecular Weight (g/mol) | 76.09 |
| MDL Number | MFCD00008495 |
| InChI Key | NKDDWNXOKDWJAK-UHFFFAOYSA-N |
| Synonym | methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether |
| PubChem CID | 8020 |
| ChEBI | CHEBI:48341 |
| IUPAC Name | dimethoxymethane |
| SMILES | COCOC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC115560010
|
Thermo Scientific Chemicals
115560010 |
1 L | Glass bottle |
chaque for $130.60
caisse de 4 chaque for $0.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC115560025
|
Thermo Scientific Chemicals
115560025 |
2.5 L | Glass bottle |
chaque for $261.65
caisse de 4 chaque for $0.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 109-87-5 | |
| 76.09 | |
| NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
| 8020 | |
| dimethoxymethane |
| C3H8O2 | |
| MFCD00008495 | |
| methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
| CHEBI:48341 | |
| COCOC |
Spécifications
| 109-87-5 | |
| Colorless | |
| 41.0°C to 42.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3525 to 1.3545 | |
| MFCD00008495 | |
| 01,671; 02,271; 06,374; 07,115 | |
| 15, 6086 | |
| Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene | |
| COCOC | |
| 76.09 | |
| 3.25 mPa.s (20°C) | |
| 76.09 | |
| Liquid |
| -105.0°C | |
| 0.8600g/mL | |
| −18°C | |
| 99.5% min. (GC) | |
| C3H8O2 | |
| CH2(OCH3)2 | |
| 1 L | |
| 0.86 | |
| methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
| NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
| dimethoxymethane | |
| 8020 | |
| CHEBI:48341 | |
| 99.5+% | |
| Dimethoxymethane |
Sécurité et manipulation
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-714-2
RUO – Research Use Only