missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Shoe Covers

Serged seam shoe covers feature an anti-skid PVC sole and are suitable for ISO 6 (Class 1000) controlled environments.

$428.75 - $464.54

Specifications

Material Tyvek
Latex Free Yes
Color White
Disposable Single-use
Closure Type Elastic Ankle
View More Specs

Products 4
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Size Length (English) Length (Metric) Price Quantity  
29-636-1227
View Documents
DuPont™
IC461SWHSM03000B
Small 10 in. 25.4 cm
Case of 300 Each for $428.75
Only null left
 
29-636-1228
View Documents
DuPont™
IC461SWHMD03000B
Medium 11.12 in. 28.24 cm
Case of 300 Each for $428.75
Only null left
 
17-988-105
View Documents
DuPont™
IC461SWHLG03000B
Large 14.25 in. 36.19 cm
Case of 300 Each for $464.54
Only null left
 
17-988-106
View Documents
DuPont™
IC461SWHXL03000B
X-Large 15.75 in. 40 cm
Case of 300 Each for $464.54
Only null left
 
Description

Description

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • 5 in. shoe cover with elastic top
  • Elastic at the toe for a more secure fit
  • PVC soles provides enhanced skid-resistance and durability
  • Bulk-packaged
Specifications

Specifications

Tyvek
White
Elastic Ankle
Serged
ASTM D3776, ASTM F2101, ASTM D774, ASTM D5034, ASTM D257, AATCC 127, 16 CFR 1610 Class 1
13.33 cm
Yes
Single-use
Anti-skid
5.4 mil
5.25 in.
Class 1000 (ISO 6)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'