missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Gel-Loading Tips, 1-200μL

An ideal choice for accurately and easily loading wells in gel electrophoresis

$146.21 - $380.25

Specifications

Volume (Metric) 1 to 200 μL
Color Natural
Length (English) 2.75 in.
Clean Claims DNase, RNase Free
For Use With (Application) For DNA sequencing or loading electrophoresis gels
View More Specs

Products 5
Catalog Number Mfr. No. Sterility Packaging Price Quantity  
Catalog Number Mfr. No. Sterility Packaging Price Quantity  
02-707-181 Edge
View Documents
N/A Non-sterile BP
Pack of 1000 Each for $146.21
Case of 10 Pack for $1,462.10
Only null left
Edge
 
02-707-138
View Documents
N/A Non-sterile HR, 10 x 96
Pack of 960 Each for $332.89
Case of 5 Pack for $1,664.45
Only null left
 
02-707-139
View Documents
N/A Sterile HRS, 10 x 96
Pack of 960 Each for $380.25
Case of 5 Pack for $1,901.25
Only null left
 
02-707-182
View Documents
N/A Non-sterile LR, 5 x 204
Pack of 1020 Each for $249.54
Case of 5 Pack for $1,247.70
Only null left
 
02-707-81
View Documents
N/A Sterile LRS, 5 x 204
Pack of 1020 Each for $290.59
Case of 5 Pack for $1,452.95
Only null left
 
Description

Description

Use in gel electrophoresis, to accurately and easily load wells.

  • Round gel-loading tip
  • Easily load gels 0.6mm
  • Applications include Agarose DNA gels, 2-D and SDS PAGE, Capillary gels and Sequencing
  • Certified by independent laboratory

Specifications

Specifications

1 to 200 μL
2.75 in.
For DNA sequencing or loading electrophoresis gels
Polypropylene
Natural
DNase, RNase Free
Gel Loading, Round
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'