Fisherbrand™ Magnetic Stir Bar Six Packs
These magnetic stirring bars are the most commonly used shape.
$16.31 - $39.17
Specifications
Autoclavable | Autoclavable |
---|---|
Coating | PTFE |
Material | Alnico Magnet |
Quantity | 6 Pk. |
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | ||
1451351SIX
|
N/A | 0.375 in. | 9.5 mm | 1 in. | 25.4 mm |
Pack of 6 Each for $26.03
|
|
||
1451357SIX
|
N/A | 0.19 in. | 4.5 mm | 0.44 in. | 12 mm |
Pack of 6 Each for $17.97
|
|
||
1451358SIX
|
N/A | 0.31 in. | 8 mm | 0.5 in. | 12.7 mm |
Pack of 6 Each for $16.31
|
|
||
1451359SIX
|
N/A | 0.31 in. | 8 mm | 1 in. | 25.4 mm |
Pack of 6 Each for $20.21
|
|
||
1451360SIX
|
N/A | 0.31 in. | 8 mm | 1.5 in. | 38.1 mm |
Pack of 6 Each for $25.52
|
|
||
1451361SIX
|
Fisherbrand™
800371119 |
0.31 in. | 8 mm | 2 in. | 50.8 mm |
Pack of 6 Each for $32.52
|
|
||
1451363SIX
|
N/A | 0.079 in. | 2 mm | 0.25 in. | 7 mm |
Pack of 6 Each for $29.93
|
|
||
1451364SIX
|
N/A | 0.059 in. | 1.5 mm | 0.31 in. | 8 mm |
Pack of 6 Each for $29.52
|
|
||
1451366SIX
|
Fisherbrand™
800371120 |
0.375 in. | 9.5 mm | 2 in. | 50.8 mm |
Pack of 6 Each for $39.17
|
|
||
1451365SIX
|
N/A | 0.118 in. | 3 mm | 0.39 in. | 10 mm |
Pack of 6 Each for $19.79
|
|
||
Description
These magnetic stirring bars are the most commonly used shape.
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
- Autoclavable
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
Specifications
Autoclavable | |
PTFE | |
Alnico Magnet | |
6 Pk. |