missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Two-Hole Rubber Stoppers
Assorted sizes 00 to 15
$80.04 - $108.93
Specifications
Color | Black |
---|---|
Material | Natural Rubber |
Product Type | Tapered Stopper (2 holes) |
For Use With (Application) | General purpose use |
Total Positions | 2 |
Catalog Number | Mfr. No. | Length (Metric) | Size | Stopper Number | Diameter (Metric) Top | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Size | Stopper Number | Diameter (Metric) Top | Price | Quantity | ||
14-140S
|
N/A | 3.9 cm | 15 | 15 | 10.3 cm |
Each for $86.82
Case of 7 Each for $607.74
|
|
||
14-140A
|
N/A | 2.5 cm | 0 | 0 | 1.5 cm |
Pack of 113 Each for $81.07
Case of 5 Pack for $405.35
|
|
||
14-140B
|
N/A | 2.5 cm | 0 | 0 | 1.7 cm |
Pack of 82 Each for $80.04
Case of 5 Pack for $400.20
|
|
||
14-140D
|
N/A | 2.5 cm | 2 | 2 | 2 cm |
Pack of 57 Each for $98.79
Case of 5 Pack for $493.95
|
|
||
14-140J
|
N/A | 2.5 cm | 7 | 7 | 3.7 cm |
Pack of 16 Each for $93.07
Case of 5 Pack for $465.35
|
|
||
14-140K
|
N/A | 2.5 cm | 8 | 8 | 4.1 cm |
Pack of 13 Each for $89.36
Case of 5 Pack for $446.80
|
|
||
14-140M
|
N/A | 2.5 cm | 10 | 10 | 5 cm |
Pack of 8 Each for $108.93
Case of 5 Pack for $544.65
|
|
||
14-140P
|
N/A | 2.5 cm | 12 | 12 | 6.4 cm |
Pack of 5 Each for $104.61
Case of 5 Pack for $523.05
|
|
||
Description
Assorted sizes 00 to 15
- Hole size: 3 to 5mm
†The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).
Specifications
Black | |
Tapered Stopper (2 holes) | |
2 |
Natural Rubber | |
General purpose use |