missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Glass Erlenmeyer Flask

Glass Erlenmeyer flask with screw thread neck in standard socket size. Made with high quality borosilicate 3.3 glass, extremely durable, with a low temperature gradient and superior chemical resistance. Flask connects firmly to other glassware with the help of threaded joints.

$30.44 - $74.53

Specifications

Closure Color White
Color Clear
Description Screw thread
Material Borosilicate Glass (3.3)
For Use With (Application) Heating / Holding Liquids
View More Specs

Products 10
Catalog Number Mfr. No. Capacity (Metric) Thread Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Thread Size Price Quantity  
S40842
View Documents
Eisco™
CH01006A
50 mL 14/23
Each for $30.44
 
S40843
View Documents
Eisco™
CH01006B
50 mL 19/26
Each for $33.79
 
S40844
View Documents
Eisco™
CH01006C
100 mL 14/23
Each for $33.79
 
S40845
View Documents
Eisco™
CH01006D
100 mL 19/26
Each for $33.79
 
S40846
View Documents
Eisco™
CH01006E
100 mL 24/29
Each for $33.88
 
S40847
View Documents
Eisco™
CH01006F
250 mL 19/26
Each for $36.71
 
S40848
View Documents
Eisco™
CH01006G
250 mL 24/29
Each for $36.76
 
S40850
View Documents
Eisco™
CH01006H
500 mL 19/26
Each for $51.56
 
S40852
View Documents
Eisco™
CH01006I
500 mL 24/29
Each for $51.56
 
S40855
View Documents
Eisco™
CH01006J
1000 mL 24/29
Each for $74.53
 
Description

Description

  • Made from high quality borosilicate 3.3 glass
  • Erlenmeyer style flasks with screw thread necks
  • Extremely durable
  • Low temperature gradient
  • Superior chemical resistance
  • Connect firmly to other glassware with the help of threaded joints
  • Threaded joints provide additional safety in every lab
  • Autoclavable
  • Warranty and Services

    1 year limited manufacturer warranty

    Specifications

    Specifications

    White
    Screw thread
    Heating / Holding Liquids
    Autoclavable
    Yes
    Non-sterile
    Screw
    1
    Clear
    Borosilicate Glass (3.3)
    Conical
    Reusable
    Narrow
    Flat
    Non-treated
    Videos
    SDS
    Documents

    Documents