Eisco™ Low Form Borosilicate Glass Beakers
These low form beakers are made by hand from high quality borosilicate 3.3 glass. They are extremely durable, have low temperature gradients, and superior chemical durability.
$10.15 - $279.94
Specifications
Description | Chemical durability |
---|---|
Material | Borosilicate Glass |
Type | Glass Beakers |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | ||||||
S39780
|
Eisco™
CH0126A |
5 mL |
Each for $14.41
|
|
||||||
S40887
|
Eisco™
CH0126F |
150 mL |
Each for $10.15
|
|
||||||
S27520
|
Eisco™
CH0126J8 |
800 mL |
Each for $34.50
|
|
||||||
S39781
|
Eisco™
CH0126B |
10 mL |
Each for $18.03
|
|
||||||
S40888
|
Eisco™
CH0126J |
600 mL |
Each for $18.79
|
|
||||||
S39782
|
Eisco™
CH0126C |
25 mL |
Each for $21.56
|
|
||||||
S40889
|
Eisco™
CH0126N |
5000 mL |
Each for $279.94
|
|
||||||
S15437
|
Eisco™
CH0126D |
50 mL |
Each for $11.09
|
|
||||||
S15438
|
Eisco™
CH0126E |
100 mL |
Each for $13.29
|
|
||||||
S15439
|
Eisco™
CH0126G |
250 mL |
Each for $13.29
|
|
||||||
S15440
|
Eisco™
CH0126H |
400 mL |
Each for $18.35
|
|
||||||
S15441
|
Eisco™
CH0126I |
500 mL |
Each for $22.06
|
|
||||||
S15442
|
Eisco™
CH0126K |
1000 mL |
Each for $40.26
|
|
||||||
S15443
|
Eisco™
CH0126L |
2000 mL |
Each for $68.44
|
|
||||||
S39783
|
Eisco™
CH0126M |
3000 mL |
Each for $172.91
|
|
||||||
Description
- Made from high quality borosilicate 3.3 glass
- White graduations for easy visibility (not available for 5 mL and 10 mL sizes)
- Low temperature gradient - can withstand high temperatures and thermal shocks
- Superior chemical durability
- Low form
- Autoclavable at 121°C
- In compliance with DIN 12231 and ISO 3819 standards
Specifications
Chemical durability | |
Glass Beakers | |
Autoclavable | |
Low Form |
Borosilicate Glass | |
Non-sterile | |
Reusable |