missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals
$43.09 - $3,932.01
Chemical Identifiers
CAS | 57-09-0 |
---|---|
Molecular Formula | C19H42BrN |
Molecular Weight (g/mol) | 364.46 |
MDL Number | MFCD00011772 |
InChI Key | LZZYPRNAOMGNLH-UHFFFAOYSA-M |
PubChem CID | 5974 |
ChEBI | CHEBI:3567 |
SMILES | [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC227160100
|
Thermo Scientific Chemicals
227160100 |
10 g | Glass Bottle |
N/A
|
|
||||
AC227160250
|
Thermo Scientific Chemicals
227160250 |
25 g | Plastic Bottle |
N/A
|
|
||||
AC227161000
|
Thermo Scientific Chemicals
227161000 |
100 g | Plastic Bottle |
N/A
|
|
||||
AC227165000
|
Thermo Scientific Chemicals
227165000 |
500 g | Plastic Bottle |
N/A
|
|
||||
AC227160025
|
Thermo Scientific Chemicals
227160025 |
2.5 kg | Plastic Drum |
N/A
|
|
||||
AC227160101
|
Thermo Scientific Chemicals
227160101 |
10 kg | Plastic drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
57-09-0 | |
364.46 | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
CHEBI:3567 |
C19H42BrN | |
MFCD00011772 | |
5974 | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Specifications
White | |
Crystalline Powder | |
>230°C | |
C19H42BrN | |
MFCD00011772 | |
04, IV, 819 | |
15, 2024 | |
Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
hexadecyltrimethylazanium bromide | |
5974 | |
364.45 | |
Authentic | |
Hexadecyltrimethylammonium bromide |
57-09-0 | |
230.0°C | |
99+% | |
CH3(CH2)15N(CH3)3Br | |
10 g | |
15,77 | |
Cetrimonium bromide, Cetyltrimethylammonium bromide, CTABr | |
244°C | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
364.46 | |
CHEBI:3567 | |
≥99% | |
Glass Bottle |
Safety and Handling
Warning
RUO – Research Use Only