missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Keystone Safety KEYGUARD™ Microporous Coveralls, Elastic Wrists, Attached Hood and Boots
$306.68 - $326.01
Specifications
Gender | Unisex |
---|---|
Material | Polyolefin |
Color | White |
Disposable | Single-use |
Closure Type | Front Zipper |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-055-549
|
Keystone Safety
CVLKGBXL |
X-Large |
Case of 25 Each for $326.01
|
|
|||||
19-055-546
|
Keystone Safety
CVLKGBSM |
Small |
Case of 25 Each for $306.68
|
|
|||||
19-055-548
|
Keystone Safety
CVLKGBLG |
Large |
Case of 25 Each for $326.01
|
|
|||||
19-055-551
|
Keystone Safety
CVLKGB3X |
3X-Large |
Case of 25 Each for $310.93
|
|
|||||
19-055-550
|
Keystone Safety
CVLKGB2X |
2X-Large |
Case of 25 Each for $306.68
|
|
|||||
19-055-552
|
Keystone Safety
CVLKGB4X |
4X-Large |
Case of 25 Each for $315.01
|
|
|||||
19-055-553
|
Keystone Safety
CVLKGB5X |
5X-Large |
Case of 25 Each for $318.92
|
|
|||||
Specifications
Unisex | |
White | |
Front Zipper | |
Flame | |
Yes |
Polyolefin | |
Single-use | |
Serged | |
ANSI | |
Manufacturing, food service, industrial, medical, clean room, pharmaceutical |