DWK Life Sciences Kimble™ KIMAX™ Class B Mixing Cylinders
Calibrated To Contain
$301.35 - $1687.35
Specifications
Material | Borosilicate Glass |
---|---|
Class | B |
Closure Material | Borosilicate Glass |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | |||||
08-566-11B
|
DWK Life Sciences
2004025 |
25 mL | 0.5 mL |
Pack of 4 Each for $393.56
Case of 6 Pack for $2,361.36
|
|
|||||
08-566-11C
|
DWK Life Sciences
2004050 |
50 mL | 1 mL |
Pack of 4 Each for $545.29
Case of 6 Pack for $3,271.74
|
|
|||||
08-566-11E
|
DWK Life Sciences
20040250 |
250 mL | 2 mL |
Pack of 2 Each for $378.18
Case of 4 Pack for $1,512.72
|
|
|||||
08-566-11A
|
DWK Life Sciences
2004010 |
10 mL | 0.2 mL |
Pack of 4 Each for $320.50
Case of 6 Pack for $1,923.00
|
|
|||||
08-566-11D
|
DWK Life Sciences
20040100 |
100 mL | 1 mL |
Pack of 4 Each for $628.97
Case of 6 Pack for $3,773.82
|
|
|||||
08-566-11G
|
DWK Life Sciences
200401000 |
1000 mL | 10 mL |
Case of 4 Each for $1,687.35
|
|
|||||
08-566-11F
|
DWK Life Sciences
20040500 |
500 mL | 5 mL |
Each for $301.35
Case of 6 Each for $1,808.10
|
|
|||||
08-566-11H
|
DWK Life Sciences
200402000 |
2000 mL | 20 mL |
Case of 2 Each for $1,252.03
|
|
|||||
Description
- Single metric blue ceramic-enamel scale
- Heights shown in ordering table do not include stopper
- The 10 and 25mL sizes are short form for extra stability
Compliance
Designed from ASTM™ E 1272, Class B, Style 2 specifications; the 250mL size meets ASTM D 460 requirements; the 500mL size meets ASTM D 244 requirements
Specifications
Borosilicate Glass | |
B | |
Borosilicate Glass | |
Autoclavable |