MilliporeSigma™ Immobilon Western HRP Chemiluminescence Substrates
Premixed, ready-to-use chemiluminescent reagents for detection of HRP-based Westerns
$199.03 - $577.00
Specifications
Product Type HRP Substrate
For Use With (Application) Dot/Slot Blot
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Quantity Sensitivity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Sensitivity Price Quantity  
WBLUF0500
View Documents
MilliporeSigma™
WBLUF0500
HRP Substrate 500 mL ∼400 fg
Each for $577.00
WBLUR0500
View Documents
MilliporeSigma™
WBLUR0500
HRP Substrate 500 mL ∼1 to 3 pg
Each for $417.00
WBLUF0100A
View Documents
MilliporeSigma™
WBLUF0100
HRP Substrate 100 mL ∼400 fg
Each for $199.03
WBLUC0500
View Documents
MilliporeSigma™
WBLUC0500
HRP Substrate 500 mL ∼6 pg
Each for $292.99
Description
Description
Horseradish peroxidase (HRP) catalyzes the oxidation of luminol in the presence of peroxide, releasing photons (chemiluminescence) that are captured on x-ray films or by CCD cameras. Most HRP substrates are plagued by quickly decaying signal and short shelf life, but Luminata substrates are formulated differently to include stabilizing reagents.
  • Consistent results with less pipetting error
The stabilizers added to Luminata substrates:
  • Prevent spontaneous oxidation of luminol by peroxide in the absence of HRP
  • Enhance the stability of both the luminol and peroxide, ensuring that Luminata substrates are stable for one year in their premixed format
  • Stabilize, strengthen, and prolong the chemiluminescence signal once HRP starts to catalyze oxidation
Three different sensitivities for a range of detection needs:
  • Immobilon Classico premixed reagent for everyday detection needs
  • Immobilon Crescendo premixed reagent for detection in the picogram range
  • Immobilon Forte premixed reagent for detection in the femtogram range
Specifications
Specifications
HRP Substrate
Dot/Slot Blot
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'