missing translation for 'onlineSavingsMsg'
Learn More

Methyl acrylate, 99%, stabilized

CAS: 96-33-3 | C4H6O2 | 86.09 g/mol

$85.36 - $659.38

Chemical Identifiers

CAS 96-33-3
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008627
InChI Key BAPJBEWLBFYGME-UHFFFAOYSA-N
Synonym methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
PubChem CID 7294
ChEBI CHEBI:82482
IUPAC Name methyl prop-2-enoate
SMILES COC(=O)C=C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126190100
SDS View Documents
Thermo Scientific Chemicals
126190100
10 mL Glass bottle
Each for $85.63
Only null left
 
AC126195000
SDS View Documents
Thermo Scientific Chemicals
126195000
500 mL Glass bottle
Each for $85.36
Only null left
 
AC126190010
SDS View Documents
Thermo Scientific Chemicals
126190010
1 L Glass bottle
Each for $118.28
Case of 4 Each for $0.00
Only null left
 
AC126190025
SDS View Documents
Thermo Scientific Chemicals
126190025
2.5 L Glass bottle
Each for $255.34
Only null left
 
AC126190101
SDS View Documents
Thermo Scientific Chemicals
126190101
10 L Metal Drum
Each for $659.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-33-3
86.09
BAPJBEWLBFYGME-UHFFFAOYSA-N
7294
methyl prop-2-enoate
C4H6O2
MFCD00008627
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
CHEBI:82482
COC(=O)C=C

Specifications

96-33-3,150-76-5
0.9560g/mL
-3°C
98.5% min. (GC)
C4H6O2
H2C=CHCO2CH3
10 mL
05,439; 13,183; 17,183
15, 6085
Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol and ether
COC(=O)C=C
86.09
7294
CHEBI:82482
99%
Methyl acrylate, Stabilized
-75°C
80°C
Authentic
Glass bottle
1.4010 to 1.404
MFCD00008627
02, 399
0.956
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
BAPJBEWLBFYGME-UHFFFAOYSA-N
methyl prop-2-enoate
10 to 20ppm MEHQ
0.50 mPa.s (20°C)
86.09
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic if inhaled

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Ground/Bond container and receiving equipment.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi

GHS Signal Word: Danger

EINECSNumber : 202-500-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only