Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes
Enhance aseptic techniques with these externally threaded general long-term storage cryogenic tubes, ideal for research and general lab storage.
$464.24 - $2399.93
Specifications
Product Type | Cryogenic Screw Top |
---|---|
Material | Polypropylene |
Thread Style | External |
Product Line | Nalgene™ |
Alert:
Do not store in the liquid phase of nitrogen without CryoFlex tubing
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Price | Quantity | ||||
03-337-7X
|
Thermo Scientific™
50001012 |
1.0 mL | Sterile |
Case of 500 Each for $1,105.37
|
|
||||
03-337-7C
|
Thermo Scientific™
50000012 |
1.2 mL | Sterile |
Case of 500 Each for $991.02
|
|
||||
03-337-7Y
|
Thermo Scientific™
N50001020 |
1.5 mL | Sterile |
Case of 500 Each for $954.67
|
|
||||
03-337-7D
|
Thermo Scientific™
50000020 |
2.0 mL | Sterile |
Case of 500 Each for $967.55
|
|
||||
03-337-7H
|
Thermo Scientific™
50000050 |
5.0 mL | Sterile |
Case of 250 Each for $840.73
|
|
||||
03-337-7B
|
Thermo Scientific™
50050015 |
15 mL | Sterile |
Case of 75 Each for $464.24
|
|
||||
03-337-7N
|
Thermo Scientific™
50110012 |
1.2 mL | Non-sterile |
Case of 1000 Each for $2,399.93
|
|
||||
03-337-7P
|
Thermo Scientific™
50110020 |
2.0 mL | Non-sterile |
Case of 1000 Each for $2,260.00
|
|
||||
03-337-7V
|
Thermo Scientific™
50120012 |
1.2 mL | Sterile |
Case of 1000 Each for $1,862.27
|
|
||||
03-337-7W
|
Thermo Scientific™
50120020 |
2.0 mL | Sterile |
Case of 1000 Each for $1,905.58
|
|
||||
Description
Enhance aseptic techniques with externally threaded Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes. These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to vapor phase liquid nitrogen temperatures.
- Large writing areas accommodate more written sample information and graduations for easy visual volume assessment
- Noncytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
- Certified SAL of 10-6 for the highest sterility assurance available
- Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
- System 100 Tubes offer a more dense storage option (10x10) to save precious storage space
- Self standing tubes for ease of use on the benchtop
- Blank, nonsterile and uncapped tubes available in bulk for less demanding applications
Specifications
Cryogenic Screw Top | |
Polypropylene | |
External | |
Nalgene™ |